missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bezafibrate, 98+%, Thermo Scientific Chemicals

A peroxisome proliferator-activated receptor agonist and hypolipidemic agent

$48.52 - $291.60

Chemical Identifiers

CAS 41859-67-0
Molecular Formula C19H20ClNO4
Molecular Weight (g/mol) 361.82
MDL Number MFCD00078970
InChI Key IIBYAHWJQTYFKB-UHFFFAOYSA-N
Synonym bezafibrate, bezalip, cedur, bezafibrat, befizal, bezafibrato, bezafibratum, sklerofibrat, azufibrat, difaterol
PubChem CID 39042
ChEBI CHEBI:47612
IUPAC Name 2-(4-{2-[(4-chlorophenyl)formamido]ethyl}phenoxy)-2-methylpropanoic acid
SMILES CC(C)(OC1=CC=C(CCNC(=O)C2=CC=C(Cl)C=C2)C=C1)C(O)=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ61412ME
View Documents
Thermo Scientific Chemicals
J61412ME
500 mg N/A
 
AAJ6141203
View Documents
Thermo Scientific Chemicals
J6141203
1 g N/A
 
AAJ6141206
View Documents
Thermo Scientific Chemicals
J6141206
5 g N/A
 
Description

Description

A peroxisome proliferator-activated receptor agonist and hypolipidemic agent

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

41859-67-0
361.82
IIBYAHWJQTYFKB-UHFFFAOYSA-N
39042
2-(4-{2-[(4-chlorophenyl)formamido]ethyl}phenoxy)-2-methylpropanoic acid
C19H20ClNO4
MFCD00078970
bezafibrate, bezalip, cedur, bezafibrat, befizal, bezafibrato, bezafibratum, sklerofibrat, azufibrat, difaterol
CHEBI:47612
CC(C)(OC1=CC=C(CCNC(=O)C2=CC=C(Cl)C=C2)C=C1)C(O)=O

Specifications

41859-67-0
White
MFCD00078970
14,1195
IIBYAHWJQTYFKB-UHFFFAOYSA-N
2-(4-{2-[(4-chlorophenyl)formamido]ethyl}phenoxy)-2-methylpropanoic acid
39042
361.82
Powder
183°C to 186°C
C19H20ClNO4
500 mg
bezafibrate, bezalip, cedur, bezafibrat, befizal, bezafibrato, bezafibratum, sklerofibrat, azufibrat, difaterol
CC(C)(OC1=CC=C(CCNC(=O)C2=CC=C(Cl)C=C2)C=C1)C(O)=O
361.82
CHEBI:47612
≥98%
Bezafibrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

P264b-P270-P301+P312-P330-P501c

H302

EINECSNumber : 255-567-9

RTECSNumber : UE8755000

TSCA : No

Recommended Storage : Store at -20°C

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only