Abbott™ BinaxNOW™ Legionella Urinary Antigen Test Kit DFS Item
For the detection of Legionella pneumophila serogroup 1 antigen in urine specimens.
Supplier:  Abbott™ 852000
Catalog No. BNX852000
Explore available promotions
Description
Description
BinaxNOW™ Legionella Urinary Antigen Card provides accurate, rapid identification of Legionnaires’ disease caused by Legionella pneumophila serogroup 1 with an easy-to-use technology.
Rapid diagnosis of Legionnaires’ disease and early initiation of appropriate therapy has been shown to dramatically reduce morbidity and mortality. This rapid assay is used in urine samples from patients with symptoms of pneumonia. It is intended to aid in the presumptive diagnosis of Legionnaires’ disease caused by L. pneumophila serogroup 1 in conjunction with culture and other methods. 
  • Highly accurate – 95% sensitivity and specificity; treat with confidence
  • Easy-to-use – Easy to manage and train; minimizes operator error
  • Results in 15 minutes or less – Faster throughput in lab or Emergency Department, aiding in rapid, focused therapy
  • Room temperature storage – Doesn’t take up valuable refrigerator space
  • Recommended as part of the IDSA/ATS CAP Guidelines
Cost-effective part of a program to:
  • Decrease number of broad-spectrum antibiotics prescribed
  • Shorten patient wait time/hospital stay
  • Decrease need for unnecessary ancillary testing
  • Improve patient outcomes through rapid, focused therapy
Order Info
For control swabs to be used with this test, see Cat. No. BNX852010
Specifications
Specifications
Moderate
BinaxNOW™ Legionella Urinary Antigen Card
22 Tests
Urine
Qualitative detection of Legionella pneumophila serogroup 1 Antigen in Urine specimens
15 min.