Bio Plas™ Hexa-Flex™ Safety Caps
Used to cap and recap blood collection and disposable glass culture tubes
Description
Used to cap and recap blood collection and disposable glass culture tubes
- Liquid-tight seal ensures sample integrity
- Prevents cross-contamination and protects against spillage
- Assorted colors available for sample identification
- Fits six different sized tubes
Biomedical and industrial
Specifications
Glass | |
Lavender | |
10, 12, 13, 16 and 18 mm Blood Collection and Culture Tubes |