Bio Plas™ Hexa-Flex™ Safety Caps
Used to cap and recap blood collection and disposable glass culture tubes
 View more versions of this product
Catalog No. 22-228-485
Explore available promotions
Description
Description
Used to cap and recap blood collection and disposable glass culture tubes
  • Liquid-tight seal ensures sample integrity
  • Prevents cross-contamination and protects against spillage
  • Assorted colors available for sample identification
  • Fits six different sized tubes
Biomedical and industrial
Specifications
Specifications
Gray
10, 12, 13, 16 and 18 mm Blood Collection and Culture Tubes