missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bio-Serv™ Bunny Blocks™ Animal Diet, Certified

A sweet treat with a hard texture to promote good dental health

$99.73 - $381.08

Specifications

Product Type Blocks
Calories 191.5
Components Sucrose, Dextrose, Natural and Artificial Flavors, Hydrogenated Vegetable Oil, Magnesium Stearate, Food Dye
Format Pellet
Particle Shape Block
View More Specs

Products 7
Catalog Number Mfr. No. Flavor Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Flavor Packaging Quantity Price Quantity  
14726713
View Documents
Bio-Serv™
F0527325
Pineapple 25/Box
Each for $106.21
 
14726714
View Documents
Bio-Serv™
F0527425
Sweet Apple 25/Box
Each for $99.73
 
14726715
View Documents
Bio-Serv™
F0568225
Very Berry 25/Box
Each for $107.71
 
14726717
View Documents
Bio-Serv™
F05273
Pineapple 100/Box
Each for $381.08
 
14726716
View Documents
Bio-Serv™
F05781
Orange 100/Box
Each for $381.08
 
14726718
View Documents
Bio-Serv™
F05274
Sweet Apple 100/Box
Each for $352.85
 
14726719
View Documents
Bio-Serv™
F05682
Very Berry 100/Box
Each for $375.79
 
Description

Description

  • Help satisfy chewing instinct while decreasing boredom
  • 50gm fruit flavored, extra hard gnawing
  • Hard texture promotes good dental health
  • Certified (Contaminant Screened)

Certifications

Contaminant Screened

Specifications

Specifications

Blocks
Sucrose, Dextrose, Natural and Artificial Flavors, Hydrogenated Vegetable Oil, Magnesium Stearate, Food Dye
Block
Rabbit
1.75 in.
191.5
Pellet
50 g
Cool dry place
0.445 cm
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'