missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bio-Serv™ Fruit Crunchies™ Animal Diet, Certified DFS Item

A nutritional treat with a medley of flavors

Supplier:  Bio-Serv™ S057981

 View more versions of this product

Catalog No. 14726773


Explore available promotions

Description

Description

  • Nutritionally complete 190mg treat that compliments enrichment programs
  • A medley of apple, grape and orange flavors add a variety to foraging activity
  • Less labor intensive and more sanitary than perishable fruit
  • Excellent reward or foraging treat
  • Certified (Contaminant Screened)
  • Excellent source of vitamins
  • Nutritionally assayed
  • Weight: 19mg
  • Shape: pellet
  • S05798-1 is sterilized by gamma irradiation

Certifications

Contaminant screened

Specifications

Specifications

Fruit Crunchies
Casein, Dextrose, Sucrose, Cellulose, Corn Starch, Corn Oil, Corn Syrup, Mineral Mix, Choline Bitartrate, Vitamin Mix, Natural and Artificial Flavors, Calcium Silicate, Magnesium Stearate, L-Cystine, Ascorbic Acid, Food Dye
Pellet
Pellet
Sterile
0.66
Fruit Crunchies
1 kg/bag
1000 g
Cool dry place
Videos
SDS
Documents

Documents

Promotions

Promotions