missing translation for 'onlineSavingsMsg'
Learn More

Bio-Serv™ K-9 Beef Treats™, Certified DFS Item

Roast beef flavored treat has been nutritionally assayed and can be used as an appetite stimulant

Supplier:  Bio-Serv™ F05411

Catalog No. 14-726-405


Only null left
Explore available promotions

Description

Description

  • Nutritionally fortified 5g (0.2 oz.) grain-based treat
  • Highly palatable — great appetite stimulant
  • Certified (contaminant screened)
  • Hearty roast beef flavor
  • Nutritionally assayed
  • Weight: 5g (0.2 oz)
  • Shape: wafer
  • 1000/Box
Specifications

Specifications

K-9 Beef Treats
Hearty Roast Beef
Beef
1 in.
Ground Corn, Banana Flakes, Dehulled Soybean Meal, Corn Gluten Meal, Ground Wheat, Corn Gluten Feed, Fish Meal, Dehydrated Alfalfa Meal, Casein, Dried Whey, Sucrose, Fructose, Dextrose, Dried Beet Pulp, Soybean Oil, Porcine Animal Fat (preserved with BHA), Dried Brewers Yeast, Mineral Mix, Vitamin Mix, Beef Flavor, Magnesium Stearate, Calcium Silicate DL-Methionine, Choline Chloride, Sodium Propionate, Ascorbic Acid, Kaolin
1000
Cool dry place
2.5 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions