Bio-Serv™ Liquid Diet Feeding Tubes
Designed specifically for liquid diets—eliminates clogging that occurs with sipper tubes
Supplier:  Bio-Serv™ BIO9010
 View more versions of this product
Catalog No. 14727057
Explore available promotions
Requires : For use with stainless steel tube holders (Mfr. Nos. 9015 and 9016)
Description
Description
    • Made of borosilicate glass with autoclavable screw cap
    • Specifically designed to dispense Liquid Diets
    • Eliminates clogging that occurs with sipper tubes
    • Easy to read calibrations for accurate food intake
    • For use with our stainless steel tube holders
    • Designed for use with most cages
    • Autoclavable
    • Cagewasher safe
Specifications
Specifications
Liquid Diet Feeding Tube
100 mL
For liquid diet feeding
Autoclavable
Borosilicate Glass