Bio-Serv™ Rat Retreats™ Rodent Enrichment Device, Certified
Allows for sheltering and gnawing opportunities
Supplier:  Bio-Serv™ K3246
 View more versions of this product
Catalog No. 14726563
Explore available promotions
Description
Description
  • Flat top allows for increased floor space
  • Large flat sides prevent rolling
  • Made of high temperature polycarbonate
  • Autoclavable
  • Cagewasher safe
  • Certified (Contaminant screened)
Specifications
Specifications
Rat Retreat
Amber
Allows for sheltering and gnawing opportunities
6 in.
3.75 in.
3.75 in.
Autoclavable
Polycarbonate
6 x 3.75 x 3.75 in. (15.2 x 9.5 x 9.5 cm)
15.2 cm
9.5 cm
9.5 cm