Ansell BioClean™ BAFS 601 Anti-fog Goggle Spray
Alcohol-free, with airless pump action for use with autoclavable goggles.
Supplier:  Ansell BAFS601
Catalog No. 19-668-261
Explore available promotions
Description
Description
  • Anti-fog effect: For less obstruction when using autoclavable goggles.
  • Airless pump action: Quick and easy application.
  • Manufactured at an ISO 9001-certified facility: For assured quality.
Specifications
Specifications
Anti-fog
Cleaning Solution
BAFS
10/Pk.
Proprietary Formula
Not Autoclavable
Clear
100/Cs.
60 mL
Universal
Single-use