Ansell BioClean™ Nitrile Cleanroom RABS/Isolator Gloves
BioClean™ Nitrile Clean RABS/Isolator Gloves are manufactured from nitrile with incredibly low levels of particles and excellent ESD properties. Designed for use in injectable manufacturing and pharmaceutical production areas, our CGL series of gloves are cleanroom processed and packed.
$235.05 - $493.23
Specifications
Color White
Latex Free Yes
Size 9.75
Length (English) 33 in.
Length (Metric) 84 cm
View More Specs
Products 3
Catalog Number Mfr. No. Port Size (English) Port Size (Metric) Price Quantity  
Catalog Number Mfr. No. Port Size (English) Port Size (Metric) Price Quantity  
19-010-355
View Documents
Ansell
CGL20NIT59
8 to 10 in. 20.3 to 25.4 cm
Each for $235.05
19-010-356
View Documents
Ansell
CGL10NIT59
10 to 12 in. 25.4 to 30.5 cm
Each for $493.23
19-010-357
View Documents
Ansell
CGL36NIT59
12 to 14 in. 30.5 to 35.6 cm
Each for $287.01
Description
Description
  • Tested against ASTM D6978-05 for handling chemo drugs.
  • 100% water leak tested.
  • Ultra-clean surface ensures product protection.
  • Specially designed to minimize hand fatigue.
  • Autoclave compatible.
  • ISO 4 Cleanroom laundered and packaged.
  • Resistant to VHP, IPA, and disinfectants.
Specifications
Specifications
White
9.75
84 cm
Yes
33 in.
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'