missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Heathrow Scientific™ Biohazard Disposal Bags

Biohazard Disposal Bags for personal or general lab use, multiple sizes available to suit all needs.

Supplier:  Heathrow Scientific™ HS10044H

 View more versions of this product

Catalog No. 03-448-270


Explore available promotions

Description

Description

  • Biohazard collection bag
  • Biohazard warning symbol and precautions printed in English, Spanish, and French
  • Includes an indicator patch that darkens when autoclaved
  • Autoclavable to 250°F (121°C)
  • Disposable
  • HS10044H bag holder is ONLY for use with HS10320 2 L bag
Specifications

Specifications

Bag Holder
Autoclavable
2 L
Reusable
HS10320 Biohazard Disposal Bag ONLY
Non-sterile
Shrink Wrap
3.6 x 5 x 8.4 in.
9.1 cm
12.8 cm
21.3 cm
0.16 kg
Biohazard Disposal Bag Holder for HS10320 ONLY
0.5 gal.
Red
Biohazard Disposal Bags (8 x 12 in.)
HDPE-coated Steel Wire
24 Pk.
3.6 x 5 in.
3.6 in.
5 in.
8.4 in.
0.36 lb.
Videos
SDS
Documents

Documents

Promotions

Promotions