missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Heathrow Scientific™ Biohazard Disposal Bags

Biohazard Disposal Bags for personal or general lab use, multiple sizes available to suit all needs.

Supplier:  Heathrow Scientific™ HS10321

 View more versions of this product

Catalog No. 03-448-272


Explore available promotions

Description

Description

  • Biohazard collection bag
  • Biohazard warning symbol and precautions printed in English, Spanish, and French
  • Includes an indicator patch that darkens when autoclaved
  • Autoclavable to 250°F (121°C)
  • Disposable
  • HS10044H bag holder is ONLY for use with HS10320 2 L bag
Specifications

Specifications

Benchtop Biohazard Disposal Bag
Autoclavable
9 L
Single-use
Autoclave
Non-sterile
200 Pk.
63 mil
14 x 19 x 0.06 in.
35.6 cm
48.3 cm
0.15 cm
0.01 kg
Biohazard Disposal Bags
2.4 gal.
Red
Biohazard Disposal
Polypropylene
1.6 mm
Box
14 x 19 in.
14 in.
19 in.
0.06 in.
0.03 lb.
Videos
SDS
Documents

Documents

Promotions

Promotions