Biomedical Polymers Translucent Multipurpose Containers
High-quality, non-graduated stackable containers with tight-fitting lids
Supplier:  Biomedical Polymers BMPCN804
 View more versions of this product
Catalog No. 22-544-225
Explore available promotions
Description
Description
  • Ideal for storing biologically hazardous samples for terminal autoclaving before disposal
  • Molded from either high-density polyethylene or polypropylene, containers are inert to most chemicals
  • Resistant to freezing and boiling
  • Easy to write on with either pen or pencil
Specifications
Specifications
8 oz., 237 mL
8 oz.
Polyethylene
237 mL
Translucent
Translucent