Biomedical Polymers Translucent Multipurpose Containers
High-quality, non-graduated stackable containers with tight-fitting lids
Supplier: Biomedical Polymers BMPCN806
Description
- Ideal for storing biologically hazardous samples for terminal autoclaving before disposal
- Molded from either high-density polyethylene or polypropylene, containers are inert to most chemicals
- Resistant to freezing and boiling
- Easy to write on with either pen or pencil
Specifications
16 oz., 473 mL | |
16 oz. | |
Polyethylene |
473 mL | |
Translucent | |
Translucent |