FEATHER Biopsy Punch
Feather Biopsy Punch offers excellent sharpness by ultra-precision sharpening technology, which originates in surgical blade manufacturing.
Supplier:  FEATHER FBP50
 View more versions of this product
Catalog No. 361046714
Explore available promotions
Description
Description
  • Made in Japan
  • Packaged with safety cover
  • Anti-roll hexagon design
  • Completely-sealed package by special film
  • 20/Box
  • Sterilized by Gamma radiation
  • Single use only
Specifications
Specifications
Biopsy Punch
Single-use
Sterile
5.0 mm
Product Suggestions
Product Suggestions