Learn More
MilliporeSigma™ Supelco™ BIOshell™ IgG C18 HPLC Column, 1,000 Å, 2.7 μm Particle Size
BIOshell™ IgG 1,000 Å C18 is a high-speed, high-performance liquid chromatography column based on a new wide-pore (1,000 Å) Fused-Core™ particle design.
$1076.25 - $2078.55
Specifications
Particle Size 2.7 μm
pH 1 to 8
Pore Size 1000 Å
Surface Area 22 m2/g
Stationary Phase C18 (octadecyl)
View More Specs
Products 12
Catalog Number Mfr. No. Length (Metric) Diameter (Metric) Price Quantity  
Catalog Number Mfr. No. Length (Metric) Diameter (Metric) Price Quantity  
111007297
View Documents
MilliporeSigma™ Supelco™
581362U
5 cm 2.1 mm
Each for $1,339.33
Only null left
111006973
View Documents
MilliporeSigma™ Supelco™
582701U
10 cm 2.1 mm
Each for $1,842.25
Only null left
111006771
View Documents
MilliporeSigma™ Supelco™
582703U
15 cm 2.1 mm
Each for $2,028.65
Only null left
111006558
View Documents
MilliporeSigma™ Supelco™
582704U
25 cm 2.1 mm
Each for $2,078.55
Only null left
111007298
View Documents
MilliporeSigma™ Supelco™
582705U
3 cm 3 mm
Each for $1,076.25
Only null left
111007299
View Documents
MilliporeSigma™ Supelco™
582706U
5 cm 3 mm
Each for $1,243.67
Only null left
111006974
View Documents
MilliporeSigma™ Supelco™
582707U
10 cm 3 mm
Each for $1,737.97
Only null left
111006772
View Documents
MilliporeSigma™ Supelco™
582708U
15 cm 3 mm
Each for $2,028.65
Only null left
111007300
View Documents
MilliporeSigma™ Supelco™
582709U
3 cm 4.6 mm
Each for $1,076.25
Only null left
111007301
View Documents
MilliporeSigma™ Supelco™
582710U
5 cm 4.6 mm
Each for $1,243.67
Only null left
111006975
View Documents
MilliporeSigma™ Supelco™
582713U
10 cm 4.6 mm
Each for $1,737.97
Only null left
111006770
View Documents
MilliporeSigma™ Supelco™
581348U
15 cm 4.6 mm
Each for $2,028.65
Only null left
Description
Description
BIOshell™ IgG 1,000 Å C18 columns contain particles that provide a thin porous shell of high-purity silica surrounding a solid silica core. This particle design exhibits very high column efficiency due to the shallow diffusion paths in the 0.5 μm thin porous shell and the small overall particle size of 2.7 μm. The sterically protected C18 bonded phase of BIOshell™ IgG 1,000 Å C18 provides a stable, reversed-phase packing that can be used for separating high molecular weight compounds, such as proteins.
Specifications
Specifications
2.7 μm
1000 Å
C18 (octadecyl)
C18 (octadecyl) Phase/Fused-Core Particle Platform
Reversed Phase
1 to 8
22 m2/g
BIOshell
Yes
Fused-Core HPLC
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'
BIOshell is a trademark of Sigma-Aldrich Co. LLC. Fused-Core is a registered trademark of Advanced Materials Technology, Inc.