missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Agilent BioTek BioStack Automated Microplate Stacker - Microplate Stacks
Storage stacks
$2016.93 - $3075.61
Specifications
| Product Type | Microplates |
|---|---|
| For Use With (Equipment) | Bio-tek Biostack automated microplate stacker, unlidded plates ≤14 mm |
| For Use With (Application) | Storage |
Description
- One set
Specifications
| Microplates | |
| Bio-tek Biostack automated microplate stacker, unlidded plates ≤14 mm | |
| Storage |