Innovatek BioStrip™ Urinalysis Reagent Strips
Innovatek BioStrip™ Urinalysis Reagent Strips provide consistent, easily interpreted results
Supplier: Innovatek 4192U10
Description
- Excellent quality with unsurpassed economy
- Popular test configurations
- Consistent results that are easily interpreted
- Long shelf life, desiccant keeps strips fresh
Specifications
Glucose, Bilirubin, Ketone, Specific Gravity, Blood, pH, Protein, Urobilinogen, Nitrite, Leukocyte | |
Urinalysis | |
BioStrip™Â U10 |
Reagent Strip | |
Desiccant | |
100 Test Strips/Bottle |