Biosynergy HemoTemp™ II Activator
Supplier: Biosynergy 9000
Description
For use with HemoTemp™ II Indicator
- Offers ideal heat source for activating the irreversible portion of the HemoTemp II Indicator (Cat. No. 22-246-194)
- Maintains a temperature uniformity within ±0.5°C
- Holds and heats up to 12 HemoTemp II Indicators; indicators can stay in the activator up to three weeks
Compliance
UL approved
Warranty
Two year
Specifications
Activating the irreversible portion of HemoTemp II indicators | |
Activator |