Agilent BioTek Absorbance Test Plate
Provides performance verification for microplate readers.
Supplier: Agilent BioTek 7260522E
Includes:
- Exact spectrophotometric calibration data for each neutral density filter at eight wavelengths: 405, 450, 490, 550, 620, 630, 690, and 750nm
- Protective case
Description
- Tests alignment, accuracy, reproducibility and linearity
- Conforms to the size and shape of a standard microplate to fit easily in a microplate reader
- Ideal for monitoring an instrument's performance in compliance with regulatory requirements
Compliance
Uses NIST-traceable neutral density glass filters and precision machined blanking- and alignment-verification holes to confirm performance specifications.
Specifications
405; 450; 490; 550; 620; 630; 690; 750 nm |