missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Agilent BioTek Cytation 5 Multi-Mode Only Reader

Cytation 5 combines automated microscopy and conventional microplate detection in a configurable, upgradable platform. For Research Use Only. Not for use in diagnostic procedures.

$57370.03 - $120880.25

Specifications

Type Multi-Mode Only Microplate Reader
Shaking Linear, Orbital, Double Orbital
CO2 Concentration Range 0 to 20%
Absorbance Range 230 to 999 nm, 1 nm increments
Accuracy Monochromator: ±2 nm; Dispenser: ±1 μL or 2%
View More Specs

Products 5
Catalog Number Mfr. No. Model Plate Type Includes Price Quantity  
Catalog Number Mfr. No. Model Plate Type Includes Price Quantity  
BTCYT5F
View Documents
Agilent BioTek
CYT5FSN
CYT5F Monochromator: 6- to 384-well plates; Filters: 6-to 1536 well plates; Imaging: 6-to 1536 well plates Filter-based fluorescence intensity/fluorescence polarization/time resolved fluorescence/luminescence N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
BTCYT5FA
View Documents
Agilent BioTek
CYT5FASN
CYT5FA Monochromator: 6- to 384-well plates; Filters: 6-to 1536 well plates; Imaging: 6-to 1536 well plates Filter-based fluorescence intensity/fluorescence polarization/time resolved fluorescence/luminescence/alpha detection N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
BTCYT5M
View Documents
Agilent BioTek
CYT5MSN
CYT5M Monochromator: 6- to 384-well plates; Filters: 6-to 1536 well plates; Imaging: 6-to 1536 well plates; Take3 Plate compatible Monochromator-based UV-Vis absorbance/fluorescence intensity/time resolved fluorescence (secondary)/luminescence N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
BTCYT5MF
View Documents
Agilent BioTek
CYT5MFSN
CYT5MF Monochromator: 6- to 384-well plates; Filters: 6-to 1536 well plates; Imaging: 6-to 1536 well plates; Take3 Plate compatible Filter-based fluorescence intensity/fluorescence polarization/time resolved fluorescence/luminescence; Monochromator-based UV-Vis absorbance/fluorescence intensity/time resolved fluorescence (secondary)/luminescence N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
BTCYT5MFA
View Documents
Agilent BioTek
CYT5MFASN
CYT5MFA Monochromator: 6- to 384-well plates; Filters: 6-to 1536 well plates; Imaging: 6-to 1536 well plates; Take3 Plate compatible Filter-based fluorescence intensity/fluorescence polarization/time resolved fluorescence/luminescence/alpha detection; Monochromator-based UV-Vis absorbance/fluorescence intensity/time resolved fluorescence (secondary)/luminescence N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Cytation 5 combines automated microscopy and conventional microplate detection in a configurable, upgradable platform. The microscopy module offers up to 60x magnification in fluorescence, brightfield, high contrast brightfield, color brightfield, and phase contrast to address many applications and workflows

The multi-mode detection modules include filter- and monochromator-based fluorescence detection, luminescence, and UV-Vis absorbance detection. Gen5 software provides complete control over all imaging and data capture, plus powerful image and data analysis.

Specifications

Specifications

Multi-Mode Only Microplate Reader
0 to 20%
Monochromator: ±2 nm; Dispenser: ±1 μL or 2%
BioStack and Third Party Automation Compatible; BioSpa 8 Automated Incubator Compatible
2 Syringe Pumps
Xenon Flash; 100 mW 680 nm laser (Alpha Detection)
4-Zone incubation to 65°C with Condensation Control (±0.2°C at 37°C)
51.4 cm
44.5 cm
36.3 kg
41.6 cm
250 W max.
Linear, Orbital, Double Orbital
230 to 999 nm, 1 nm increments
Absorbance: 0 to 4 OD; Fluorescence Intensity: 7 decades; Luminesence: >6 decades
5 to 1000 μL, 1 μL increments
End point, kinetic, spectral scanning and well area scanning
Gen5 Microplate Reader and Imager Software included; Gen5 Image+ and Image Prime software available for full image analysis; Gen5 Secure for 21 CFR Part 11 compliance (option)
20.2 in.
17.5 in.
80 lb.
16.4 in.
CE and TUV marked. RoHS compliant.
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'