Agilent BioTek BioStack Automated Microplate Stackers
BioTek BioStack microplate stacker is a compact and versatile microplate stacker, compatible with Agilent BioTek washers, dispensers, plate readers, and imaging systems.
Supplier:  Agilent BioTek BSTCK3WRSN
 View more versions of this product
Catalog No. BIOSTACK3WR
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions
Description
Description
BioTek BioStack microplate stacker is a compact and versatile microplate stacker, compatible with Agilent BioTek washers, dispensers, plate readers, and imaging systems. The rapid plate exchange speeds deliver increased throughput and enhanced efficiency and greater productivity
BioStack can accommodate assay workflows for 24- to 1536-well plates. Microscope slides can be stacked for imaging using BioTek's custom slide holders. The BioTek BioStack 4 is a delidding microplate stacker, automating the handling of lidded microplates for sensitive cell-based assays, for complete walkaway automation.
Certifications
CE and TUV marked
Compliance
RoHS compliant
Specifications
Specifications
18 to 40°C
100/240 V
20.7 in.
10-, 30- and 50-plate stacks are removable and interchangeable; 96-/384-well plates: Up to 50 plates; 1536-well plates: Up to 75 plates
One instrument integration kit; Communication cable; Operator manual; Microplate pedestal for storage stack loading and unloading; Maintenance kit
LHC v 2.12 for liquid handling instruments (optional); Gen5 2.01 for readers
11.3 kg
18.8 cm
50/60 Hz
CE and TUV marked. RoHS compliant.
52.6 cm
10 to 85%
ANSI/SBS Standard and low profile 96-, 384- and 1536-well plates, Maximum plate height 14.6 mm
<25 lb.
7.4 in.
BioStack 3 Microplate Stacker