Lonza™ BioWhittaker™ X-VIVO™ 15 Hematopoietic Serum-Free Culture Media
For adoptive immunotherapy and cellular therapy research. BioWhittaker™ X-VIVO™ 15 Hematopoietic Serum-Free Culture Media supports the proliferation of lymphocytes, including tumor infiltrating lymphocytes (TIL) and macrophages under serum-free conditions.
Supplier: Lonza™ 04418Q
Description
- Does not contain any lymphokines required for activation of human lymphocytes
- Intended for in vitro procedures
- Device Master Records (DMRs) for these products are held in an FDA Master File
- Chemically defined media optimized for specific cell types
- For further manufacturing or laboratory use
- Supports the proliferation of lymphocytes, including tumor infiltrating lymphocytes (TIL) and macrophages under serum-free conditions
- Limited by Federal (USA) law to investigational use
- These products are for research use only
- Not approved for human or veterinary use, for application to humans or animals, or for use in clinical or in vitro procedures
Specifications
2-8°C | |
Gentamicin, Phenol Red | |
1Â L |
Stem Cell Serum Free Media (SFM) | |
None |