missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Biphenyl-2-boronic acid pinacol ester, 97%, Thermo Scientific Chemicals

$96.79 - $383.84

Chemical Identifiers

CAS 914675-52-8
Molecular Formula C18H21BO2
Molecular Weight (g/mol) 280.174
MDL Number MFCD14636459
InChI Key WCXWQEUBHZKNMQ-UHFFFAOYSA-N
Synonym biphenyl-2-boronic acid pinacol ester, 2-1,1'-biphenyl-2-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-2-biphenylyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
PubChem CID 59207619
IUPAC Name 4,4,5,5-tetramethyl-2-(2-phenylphenyl)-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=CC=C2C3=CC=CC=C3
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH3232003
View Documents
Thermo Scientific Chemicals
H3232003
1 g
Each for $96.79
 
AAH3232006
View Documents
Thermo Scientific Chemicals
H3232006
5 g
Each for $383.84
 
Description

Description

Biphenyl-2-boronic acid pinacol ester, is used as a main benefits of the use of boronic esters in Suzuki-Miyaura cross-couplings- Enabling the coupling of organoboronic acids that are sensitive to hydrolytic deboronation. It includes other additional application such as a number of chiral boronic esters, such as the ones illustrated hereafter, have been used as inducers in stereoselective reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Biphenyl-2-boronic acid pinacol ester, is used as a main benefits of the use of boronic esters in Suzuki-Miyaura cross-couplings- Enabling the coupling of organoboronic acids that are sensitive to hydrolytic deboronation. It includes other additional application such as a number of chiral boronic esters, such as the ones illustrated hereafter, have been used as inducers in stereoselective reactions.

Solubility
Insoluble in water. Soluble in organic solvents.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.
Specifications

Chemical Identifiers

914675-52-8
280.174
WCXWQEUBHZKNMQ-UHFFFAOYSA-N
59207619
B1(OC(C(O1)(C)C)(C)C)C2=CC=CC=C2C3=CC=CC=C3
C18H21BO2
MFCD14636459
biphenyl-2-boronic acid pinacol ester, 2-1,1'-biphenyl-2-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-2-biphenylyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
4,4,5,5-tetramethyl-2-(2-phenylphenyl)-1,3,2-dioxaborolane

Specifications

914675-52-8
C18H21BO2
1 g
Insoluble in water. Soluble in organic solvents.
B1(OC(C(O1)(C)C)(C)C)C2=CC=CC=C2C3=CC=CC=C3
280.174
280.17
Biphenyl-2-boronic acid pinacol ester
76°C to 79°C
MFCD14636459
biphenyl-2-boronic acid pinacol ester, 2-1,1'-biphenyl-2-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-2-biphenylyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
WCXWQEUBHZKNMQ-UHFFFAOYSA-N
4,4,5,5-tetramethyl-2-(2-phenylphenyl)-1,3,2-dioxaborolane
59207619
97%
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only