Learn More
Biphenyl-2-carboxylic acid, 98%, Thermo Scientific Chemicals
$41.68 - $374.97
Chemical Identifiers
CAS | 947-84-2 |
---|---|
Molecular Formula | C13H10O2 |
Molecular Weight (g/mol) | 198.221 |
MDL Number | MFCD00002463 |
InChI Key | ILYSAKHOYBPSPC-UHFFFAOYSA-N |
Synonym | 2-biphenylcarboxylic acid, 1,1'-biphenyl-2-carboxylic acid, o-phenylbenzoic acid, biphenyl-2-carboxylic acid, biphenylcarboxylic acid, 1,1'-biphenyl carboxylic acid, diphenyl-2-carboxylic acid, phenylbenzoic acid |
PubChem CID | 70357 |
IUPAC Name | 2-phenylbenzoic acid |
SMILES | C1=CC=C(C=C1)C2=CC=CC=C2C(=O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1204906
|
Thermo Scientific Chemicals
A1204906 |
5 g |
Each for $41.68
|
|
|||||
AAA1204914
|
Thermo Scientific Chemicals
A1204914 |
25 g |
Each for $134.05
|
|
|||||
AAA1204922
|
Thermo Scientific Chemicals
A1204922 |
100 g |
Each for $374.97
|
|
|||||
Description
It is used as a pharmaceutical intermediate. he reaction of 2?-substituted biphenyl-2-carboxylic acids (where the 2?-substituent is H, CO2H, NO2, Cl, OMe, or CO2Me) with lead tetra-acetate in refluxing benzene solution, under a nitrogen atmosphere, affords 3,4-benzocoumarin as a major organic product.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used as a pharmaceutical intermediate. he reaction of 2′-substituted biphenyl-2-carboxylic acids (where the 2′-substituent is H, CO2H, NO2, Cl, OMe, or CO2Me) with lead tetra-acetate in refluxing benzene solution, under a nitrogen atmosphere, affords 3,4-benzocoumarin as a major organic product.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep away from oxidizing agents.
Chemical Identifiers
947-84-2 | |
198.221 | |
ILYSAKHOYBPSPC-UHFFFAOYSA-N | |
70357 | |
C1=CC=C(C=C1)C2=CC=CC=C2C(=O)O |
C13H10O2 | |
MFCD00002463 | |
2-biphenylcarboxylic acid, 1,1'-biphenyl-2-carboxylic acid, o-phenylbenzoic acid, biphenyl-2-carboxylic acid, biphenylcarboxylic acid, 1,1'-biphenyl carboxylic acid, diphenyl-2-carboxylic acid, phenylbenzoic acid | |
2-phenylbenzoic acid |
Specifications
947-84-2 | |
343°C to 344°C | |
C13H10O2 | |
5 g | |
2-biphenylcarboxylic acid, 1,1'-biphenyl-2-carboxylic acid, o-phenylbenzoic acid, biphenyl-2-carboxylic acid, biphenylcarboxylic acid, 1,1'-biphenyl carboxylic acid, diphenyl-2-carboxylic acid, phenylbenzoic acid | |
ILYSAKHOYBPSPC-UHFFFAOYSA-N | |
2-phenylbenzoic acid | |
70357 | |
98% |
110°C to 114°C | |
Odorless | |
MFCD00002463 | |
974075 | |
Insoluble in water. | |
C1=CC=C(C=C1)C2=CC=CC=C2C(=O)O | |
198.221 | |
198.22 | |
Biphenyl-2-carboxylic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 213-432-1
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only