missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Biphenyl-4,4'-diboronic acid bis(pinacol) ester, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H5195603
Description
Biphenyl-4,4'-diboronic acid bis(pinacol) ester is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsBiphenyl-4,4′-diboronic acid bis(pinacol) ester is used as pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
Biphenyl-4,4'-diboronic acid bis(pinacol) ester | |
C24H32B2O4 | |
1 g | |
Slightly soluble in water. | |
CC1(C)OB(OC1(C)C)C1=CC=C(C=C1)C1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1 | |
406.14 | |
406.14 |
207611-87-8 | |
MFCD08669545 | |
4,4'-bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1,1'-biphenyl, 4,4'-biphenyldiboronic acid dipinacol ester, 4,4'-biphenyldiboronic acid pinacol ester, 4,4'-bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl biphenyl, biphenyl-4,4'-diboronic acid bis pinacol ester, 4,4'-biphenyldiboronic acid bis pinacol ester, 4,4,5,5-tetramethyl-2-4'-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1,1'-biphenyl-4-yl-1,3,2-dioxaborolane, amtb257, 4,4'-bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl biphenyl | |
INNVTCAYPFZCAL-UHFFFAOYSA-N | |
4,4,5,5-tetramethyl-2-[4-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]phenyl]-1,3,2-dioxaborolane | |
10525359 | |
95% |
Chemical Identifiers
207611-87-8 | |
406.14 | |
INNVTCAYPFZCAL-UHFFFAOYSA-N | |
10525359 | |
CC1(C)OB(OC1(C)C)C1=CC=C(C=C1)C1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1 |
C24H32B2O4 | |
MFCD08669545 | |
4,4'-bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1,1'-biphenyl, 4,4'-biphenyldiboronic acid dipinacol ester, 4,4'-biphenyldiboronic acid pinacol ester, 4,4'-bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl biphenyl, biphenyl-4,4'-diboronic acid bis pinacol ester, 4,4'-biphenyldiboronic acid bis pinacol ester, 4,4,5,5-tetramethyl-2-4'-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1,1'-biphenyl-4-yl-1,3,2-dioxaborolane, amtb257, 4,4'-bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl biphenyl | |
4,4,5,5-tetramethyl-2-[4-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]phenyl]-1,3,2-dioxaborolane |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only