missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Biphenyl-4-carboxaldehyde, 98+%, Thermo Scientific Chemicals

$68.32 - $600.81

Chemical Identifiers

CAS 3218-36-8
Molecular Formula C13H10O
Molecular Weight (g/mol) 182.22
MDL Number MFCD00006947
InChI Key ISDBWOPVZKNQDW-UHFFFAOYSA-N
Synonym 4-biphenylcarboxaldehyde, 4-biphenylaldehyde, p-phenylbenzaldehyde, 1,1'-biphenyl-4-carbaldehyde, biphenyl-4-carboxaldehyde, 4-formylbiphenyl, p-biphenylylaldehyde, 1,1'-biphenyl-4-carboxaldehyde, p-biphenylaldehyde, p-biphenylcarboxaldehyde
PubChem CID 76689
IUPAC Name 4-phenylbenzaldehyde
SMILES O=CC1=CC=C(C=C1)C1=CC=CC=C1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1583906
View Documents
Thermo Scientific Chemicals
A1583906
5 g
Each for $68.32
 
AAA1583914
View Documents
Thermo Scientific Chemicals
A1583914
25 g
Each for $226.45
 
AAA1583922
View Documents
Thermo Scientific Chemicals
A1583922
100 g
Each for $600.81
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

3218-36-8
182.22
ISDBWOPVZKNQDW-UHFFFAOYSA-N
76689
O=CC1=CC=C(C=C1)C1=CC=CC=C1
C13H10O
MFCD00006947
4-biphenylcarboxaldehyde, 4-biphenylaldehyde, p-phenylbenzaldehyde, 1,1'-biphenyl-4-carbaldehyde, biphenyl-4-carboxaldehyde, 4-formylbiphenyl, p-biphenylylaldehyde, 1,1'-biphenyl-4-carboxaldehyde, p-biphenylaldehyde, p-biphenylcarboxaldehyde
4-phenylbenzaldehyde

Specifications

3218-36-8
1.107
172°C (341°F)
MFCD00006947
606693
4-biphenylcarboxaldehyde, 4-biphenylaldehyde, p-phenylbenzaldehyde, 1,1'-biphenyl-4-carbaldehyde, biphenyl-4-carboxaldehyde, 4-formylbiphenyl, p-biphenylylaldehyde, 1,1'-biphenyl-4-carboxaldehyde, p-biphenylaldehyde, p-biphenylcarboxaldehyde
O=CC1=CC=C(C=C1)C1=CC=CC=C1
182.22
182.22
Biphenyl-4-carboxaldehyde
57°C to 61°C
184°C (11 mmHg)
C13H10O
5 g
Air Sensitive
ISDBWOPVZKNQDW-UHFFFAOYSA-N
4-phenylbenzaldehyde
76689
≥98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P264b-P270-P301+P312-P330-P501c

H302

EINECSNumber : 221-742-3

RTECSNumber : DV1767000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only