missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Biphenyl-4-carboxylic acid, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1243836

 View more versions of this product

Catalog No. AAA1243836


Explore available promotions

Description

Description

Biphenyl-4-carboxylic acid is employed in the synthesis, characterization of europium, terbium complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Biphenyl-4-carboxylic acid is employed in the synthesis, characterization of europium, terbium complexes.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with bases and oxidizing agents.
Specifications

Specifications

Biphenyl-4-carboxylic acid
223°C to 227°C
C13H10O2
500 g
4-biphenylcarboxylic acid, biphenyl-4-carboxylic acid, 1,1'-biphenyl-4-carboxylic acid, p-phenylbenzoic acid, 4-carboxybiphenyl, para phenyl benzoic acid, 4-diphenylcarboxylic acid, diphenyl-4-carboxylic acid, 4-carboxy-1,1'-biphenyl
NNJMFJSKMRYHSR-UHFFFAOYSA-N
4-phenylbenzoic acid
66724
98%
92-92-2
Odorless
MFCD00002553
973519
Insoluble in water.
C1=CC=C(C=C1)C2=CC=C(C=C2)C(=O)O
198.221
198.22

Chemical Identifiers

92-92-2
198.221
NNJMFJSKMRYHSR-UHFFFAOYSA-N
66724
C1=CC=C(C=C1)C2=CC=C(C=C2)C(=O)O
C13H10O2
MFCD00002553
4-biphenylcarboxylic acid, biphenyl-4-carboxylic acid, 1,1'-biphenyl-4-carboxylic acid, p-phenylbenzoic acid, 4-carboxybiphenyl, para phenyl benzoic acid, 4-diphenylcarboxylic acid, diphenyl-4-carboxylic acid, 4-carboxy-1,1'-biphenyl
4-phenylbenzoic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 202-203-1

RTECSNumber : DV1925100

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only