missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Biphenyl, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Supplier:  SPEX CertiPrep Group LLC S500

Catalog No. SS500


Explore available promotions

Includes: Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Description

Description

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
Biphenyl92-52-41000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

Biphenyl
2mL amber prescored glass ampule
Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
MFCD00003054
C1=CC=C(C=C1)C1=CC=CC=C1
GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
7095
MeCl
92-52-4
1 mL
C12H10
ZUOUZKKEUPVFJK-UHFFFAOYSA-N
1,1'-biphenyl
154.21
CHEBI:17097

Chemical Identifiers

92-52-4
154.21
ZUOUZKKEUPVFJK-UHFFFAOYSA-N
CHEBI:17097
C1=CC=C(C=C1)C1=CC=CC=C1
C12H10
MFCD00003054
7095
1,1'-biphenyl
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

DOTInformation : Nonhazardous

Recommended Storage : Ambient

SDS
Documents

Documents

Promotions

Promotions

Product intended for laboratory use only