missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bis[1,2-bis(diphenylphosphino)ethane]palladium(0), Thermo Scientific Chemicals

$160.93 - $256.05

Chemical Identifiers

CAS 31277-98-2
Molecular Formula C52H48P4Pd
Molecular Weight (g/mol) 903.27
MDL Number MFCD00009880
InChI Key FAFGMAGIYHHRKN-UHFFFAOYSA-N
Synonym bis 1,2-bis diphenylphosphino ethane palladium 0, pd dppe 2, pd diphos 2, bis diphos palladium 0, bis 1,2-bis diphenylphosphino ethane palladium, bis diphos palladium, bis 1,2-bis diphenylphosphino ethane-palladium 0, bis 1,2-bis diphenylphosphino ethane-palladium
PubChem CID 2724231
IUPAC Name 2-diphenylphosphanylethyl(diphenyl)phosphane;palladium
SMILES [Pd].C(CP(C1=CC=CC=C1)C1=CC=CC=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C(CP(C1=CC=CC=C1)C1=CC=CC=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA3923302
View Documents
Thermo Scientific Chemicals
03923302
0.5 g
Each for $160.93
 
AA3923303
View Documents
Thermo Scientific Chemicals
03923303
1 g
Each for $256.05
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

31277-98-2
903.27
FAFGMAGIYHHRKN-UHFFFAOYSA-N
2724231
[Pd].C(CP(C1=CC=CC=C1)C1=CC=CC=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C(CP(C1=CC=CC=C1)C1=CC=CC=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
C52H48P4Pd
MFCD00009880
bis 1,2-bis diphenylphosphino ethane palladium 0, pd dppe 2, pd diphos 2, bis diphos palladium 0, bis 1,2-bis diphenylphosphino ethane palladium, bis diphos palladium, bis 1,2-bis diphenylphosphino ethane-palladium 0, bis 1,2-bis diphenylphosphino ethane-palladium
2-diphenylphosphanylethyl(diphenyl)phosphane;palladium

Specifications

31277-98-2
MFCD00009880
Air Sensitive
FAFGMAGIYHHRKN-UHFFFAOYSA-N
2-diphenylphosphanylethyl(diphenyl)phosphane;palladium
2724231
Powder
C52H48P4Pd
0.5 g
bis 1,2-bis diphenylphosphino ethane palladium 0, pd dppe 2, pd diphos 2, bis diphos palladium 0, bis 1,2-bis diphenylphosphino ethane palladium, bis diphos palladium, bis 1,2-bis diphenylphosphino ethane-palladium 0, bis 1,2-bis diphenylphosphino ethane-palladium
[Pd].C(CP(C1=CC=CC=C1)C1=CC=CC=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C(CP(C1=CC=CC=C1)C1=CC=CC=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
903.27
903.25
Bis[1,2-bis(diphenylphosphino)ethane]palladium(0)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only