missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bis(2,2,6,6-tetramethyl-4-piperidyl) sebacate, 98%, Thermo Scientific Chemicals

 

Supplier:  Thermo Scientific Chemicals H5402522

Catalog No. AAH5402522


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Bis(2,2,6,6-tetramethyl-4-piperidyl) sebacate
82°C to 85°C
C28H52N2O4
100 g
bis 2,2,6,6-tetramethyl-4-piperidyl sebacate, bis 2,2,6,6-tetramethylpiperidin-4-yl decanedioate, tinuvin 770, sanol, sumisorb 577, tinuvin 770df, tinuvin 770ls, sanol 770, sanol ls 700, sanol ls 770
CC1(C)CNCC(C)(C)C1OC(=O)CCCCCCCCC(=O)OC1C(C)(C)CNCC1(C)C
480.73
480.72
52829-07-9
216°C (421°F)
MFCD00134709
UN2811
RWMYPXKVMUFMKS-UHFFFAOYSA-N
bis(2,2,6,6-tetramethylpiperidin-4-yl) decanedioate
164282
98%

Chemical Identifiers

52829-07-9
480.73
RWMYPXKVMUFMKS-UHFFFAOYSA-N
164282
CC1(C)CNCC(C)(C)C1OC(=O)CCCCCCCCC(=O)OC1C(C)(C)CNCC1(C)C
C28H52N2O4
MFCD00134709
bis 2,2,6,6-tetramethyl-4-piperidyl sebacate, bis 2,2,6,6-tetramethylpiperidin-4-yl decanedioate, tinuvin 770, sanol, sumisorb 577, tinuvin 770df, tinuvin 770ls, sanol 770, sanol ls 700, sanol ls 770
bis(2,2,6,6-tetramethylpiperidin-4-yl) decanedioate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H330-H318-H315-H335
Fatal if inhaled.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.

P280-P305+P351+P338-P310

H318

DOTInformation : Hazard Class: 9; Packaging Group: III

EINECSNumber : 258-207-9

RTECSNumber : HD8315000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only