missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Bis(2-benzamidophenyl) disulfide, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1211014
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Bis(2-benzamidophenyl) disulfide | |
140°C to 143°C | |
MFCD00043806 | |
721443 | |
ZHMIOPLMFZVSHY-UHFFFAOYSA-N | |
N-[2-[(2-benzamidophenyl)disulfanyl]phenyl]benzamide | |
67271 | |
97% |
135-57-9 | |
C26H20N2O2S2 | |
25 g | |
bis 2-benzamidophenyl disulfide, peptazin bafd, peptisant 1o, pepton 22, 2,2'-dibenzoylaminodiphenyl disulfide, bis o-benzamidophenyl disulfide, o,o'-dibenzamidodiphenyl disulfide, 2,2'-dibenzamidodiphenyl disulfide, n,n'-dithiodi-o-phenylenedibenzamide, 2',2'-dithiodibenzanilide | |
C1=CC=C(C=C1)C(=O)NC2=CC=CC=C2SSC3=CC=CC=C3NC(=O)C4=CC=CC=C4 | |
456.578 | |
456.59 |
Chemical Identifiers
135-57-9 | |
456.578 | |
ZHMIOPLMFZVSHY-UHFFFAOYSA-N | |
67271 | |
C1=CC=C(C=C1)C(=O)NC2=CC=CC=C2SSC3=CC=CC=C3NC(=O)C4=CC=CC=C4 |
C26H20N2O2S2 | |
MFCD00043806 | |
bis 2-benzamidophenyl disulfide, peptazin bafd, peptisant 1o, pepton 22, 2,2'-dibenzoylaminodiphenyl disulfide, bis o-benzamidophenyl disulfide, o,o'-dibenzamidodiphenyl disulfide, 2,2'-dibenzamidodiphenyl disulfide, n,n'-dithiodi-o-phenylenedibenzamide, 2',2'-dithiodibenzanilide | |
N-[2-[(2-benzamidophenyl)disulfanyl]phenyl]benzamide |
Safety and Handling
EINECSNumber : 205-201-9
RTECSNumber : CV8700000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only