missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bis(2-benzamidophenyl) Disulfide 98.0+%, TCI America™

$29.41 - $271.62

Chemical Identifiers

CAS 135-57-9
Molecular Formula C26H20N2O2S2
Molecular Weight (g/mol) 456.578
MDL Number MFCD00043806
InChI Key ZHMIOPLMFZVSHY-UHFFFAOYSA-N
Synonym bis 2-benzamidophenyl disulfide, peptazin bafd, peptisant 1o, pepton 22, 2,2'-dibenzoylaminodiphenyl disulfide, bis o-benzamidophenyl disulfide, o,o'-dibenzamidodiphenyl disulfide, 2,2'-dibenzamidodiphenyl disulfide, n,n'-dithiodi-o-phenylenedibenzamide, 2',2'-dithiodibenzanilide
PubChem CID 67271
IUPAC Name N-[2-[(2-benzamidophenyl)disulfanyl]phenyl]benzamide
SMILES C1=CC=C(C=C1)C(=O)NC2=CC=CC=C2SSC3=CC=CC=C3NC(=O)C4=CC=CC=C4
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
D110225G
View Documents
TCI America
D110225G
25 g
Each for $29.41
 
D1102500G
View Documents
TCI America
D1102500G
500 g
Each for $271.62
 
Description

Description

Specifications

Chemical Identifiers

135-57-9
456.578
ZHMIOPLMFZVSHY-UHFFFAOYSA-N
67271
C1=CC=C(C=C1)C(=O)NC2=CC=CC=C2SSC3=CC=CC=C3NC(=O)C4=CC=CC=C4
C26H20N2O2S2
MFCD00043806
bis 2-benzamidophenyl disulfide, peptazin bafd, peptisant 1o, pepton 22, 2,2'-dibenzoylaminodiphenyl disulfide, bis o-benzamidophenyl disulfide, o,o'-dibenzamidodiphenyl disulfide, 2,2'-dibenzamidodiphenyl disulfide, n,n'-dithiodi-o-phenylenedibenzamide, 2',2'-dithiodibenzanilide
N-[2-[(2-benzamidophenyl)disulfanyl]phenyl]benzamide

Specifications

135-57-9
C26H20N2O2S2
25 g
ZHMIOPLMFZVSHY-UHFFFAOYSA-N
N-[2-[(2-benzamidophenyl)disulfanyl]phenyl]benzamide
67271
≥98.0% (N)
Bis(2-benzamidophenyl) Disulfide
143°C
MFCD00043806
bis 2-benzamidophenyl disulfide, peptazin bafd, peptisant 1o, pepton 22, 2,2'-dibenzoylaminodiphenyl disulfide, bis o-benzamidophenyl disulfide, o,o'-dibenzamidodiphenyl disulfide, 2,2'-dibenzamidodiphenyl disulfide, n,n'-dithiodi-o-phenylenedibenzamide, 2',2'-dithiodibenzanilide
C1=CC=C(C=C1)C(=O)NC2=CC=CC=C2SSC3=CC=CC=C3NC(=O)C4=CC=CC=C4
456.578
456.58
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-2986

RTECSNumber : CV8700000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'