Learn More
Bis(2-chloroethyl) ether, 99%, Thermo Scientific Chemicals
$59.81 - $499.71
Chemical Identifiers
CAS | 111-44-4 |
---|---|
Molecular Formula | C4H8Cl2O |
Molecular Weight (g/mol) | 143.01 |
MDL Number | MFCD00000975 |
InChI Key | ZNSMNVMLTJELDZ-UHFFFAOYSA-N |
Synonym | 2-chloroethyl ether, 2,2'-dichlorodiethyl ether, bis 2-chloroethyl ether, chlorex, chloroethyl ether, dcee, dichloroether, khloreks, dichloroethyl ether |
PubChem CID | 8115 |
ChEBI | CHEBI:34573 |
IUPAC Name | 1-chloro-2-(2-chloroethoxy)ethane |
SMILES | ClCCOCCCl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1423418
|
Thermo Scientific Chemicals
A1423418 |
50 g |
Each for $59.81
|
|
|||||
AAA1423430
|
Thermo Scientific Chemicals
A1423430 |
250 g |
Each for $136.67
|
|
|||||
AAA142340B
|
Thermo Scientific Chemicals
A142340B |
1000 g |
Each for $499.71
|
|
|||||
Description
Bis(2-chloroethyl) ether is used as a solvent and soil fumigant. It may be used in the preparation of macrocyclic polyethers, dibenzo-18-crown-6 polyether and dicyclohexyl-18-crown-6 polyether.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsBis(2-chloroethyl) ether is used as a solvent and soil fumigant. It may be used in the preparation of macrocyclic polyethers, dibenzo-18-crown-6 polyether and dicyclohexyl-18-crown-6 polyether.
Solubility
Soluble in water. (1.1 g/L) at 20°C.
Notes
Special handling precautions required. View MSDS prior to purchase. MSDS are available online at www.alfa.com
Chemical Identifiers
111-44-4 | |
143.01 | |
ZNSMNVMLTJELDZ-UHFFFAOYSA-N | |
8115 | |
1-chloro-2-(2-chloroethoxy)ethane |
C4H8Cl2O | |
MFCD00000975 | |
2-chloroethyl ether, 2,2'-dichlorodiethyl ether, bis 2-chloroethyl ether, chlorex, chloroethyl ether, dcee, dichloroether, khloreks, dichloroethyl ether | |
CHEBI:34573 | |
ClCCOCCCl |
Specifications
111-44-4 | |
1.219 | |
55°C (131°F) | |
C4H8Cl2O | |
ClCH2CH2OCH2CH2Cl | |
50 g | |
605317 | |
2-chloroethyl ether, 2,2'-dichlorodiethyl ether, bis 2-chloroethyl ether, chlorex, chloroethyl ether, dcee, dichloroether, khloreks, dichloroethyl ether | |
ZNSMNVMLTJELDZ-UHFFFAOYSA-N | |
1-chloro-2-(2-chloroethoxy)ethane | |
8115 | |
143.01 | |
Bis(2-chloroethyl) ether |
-50°C | |
178°C | |
Pungent | |
1.457 | |
MFCD00000975 | |
UN1916 | |
14,3066 | |
Soluble in water. (1.1g/L) at 20°C. | |
ClCCOCCCl | |
143.01 | |
CHEBI:34573 | |
99% |
Safety and Handling
GHS H Statement
H300-H310-H330-H226-H351
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Flammable liquid and vapour.
Suspected of causing cancer.
P210-P233-P235-P240-P241-P242-P243-P260-P262-P264b-P270-P271-P280-P284-P301+P310-P303+P361+P353-P304+P340-P310-P330-P363-P370+P378q-P501c
H226-H300+H310+H330
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: 2,2'-DICHLORODIETHYL ETHER
EINECSNumber : 203-870-1
RTECSNumber : KN0875000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only