missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bis[2-(di-tert-butylphosphino)ethyl]amine, 10% w/w soln. in THF, Thermo Scientific Chemicals

$72.05 - $1020.37

Chemical Identifiers

CAS 944710-34-3
Molecular Formula C20H45NP2
Molecular Weight (g/mol) 361.54
MDL Number MFCD17014018
InChI Key MJYKYLNPIGDVEF-UHFFFAOYSA-N
Synonym bis 2-di-t-butylphosphino ethyl amine, bis 2-di-tert-butylphosphino ethyl amine, bis 2-di-tert-butylphosphanyl ethyl amine, bis 2-di-tert-butylphosphino ethyl amine w/w soln. in thf, bis 2-di-tert-butylphosphino ethyl amine solution, 10 wt. % in thf
PubChem CID 51346231
IUPAC Name 2-ditert-butylphosphanyl-N-(2-ditert-butylphosphanylethyl)ethanamine
SMILES CC(C)(C)P(CCNCCP(C(C)(C)C)C(C)(C)C)C(C)(C)C
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6049303
View Documents
Thermo Scientific Chemicals
H6049303
1 g
Each for $72.05
 
AAH6049306
View Documents
Thermo Scientific Chemicals
H6049306
5 g
Each for $245.25
 
AAH6049314
View Documents
Thermo Scientific Chemicals
H6049314
25 g
Each for $1,020.37
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
This compound is involved in the transition metal-catalyzed addition of amines to acrylic acid derivatives.

Solubility
Insoluble in water.

Notes
Air Sensitive. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

944710-34-3
361.54
MJYKYLNPIGDVEF-UHFFFAOYSA-N
51346231
CC(C)(C)P(CCNCCP(C(C)(C)C)C(C)(C)C)C(C)(C)C
C20H45NP2
MFCD17014018
bis 2-di-t-butylphosphino ethyl amine, bis 2-di-tert-butylphosphino ethyl amine, bis 2-di-tert-butylphosphanyl ethyl amine, bis 2-di-tert-butylphosphino ethyl amine w/w soln. in thf, bis 2-di-tert-butylphosphino ethyl amine solution, 10 wt. % in thf
2-ditert-butylphosphanyl-N-(2-ditert-butylphosphanylethyl)ethanamine

Specifications

944710-34-3
Unpleasant
1.418
MFCD17014018
UN1993
bis 2-di-t-butylphosphino ethyl amine, bis 2-di-tert-butylphosphino ethyl amine, bis 2-di-tert-butylphosphanyl ethyl amine, bis 2-di-tert-butylphosphino ethyl amine w/w soln. in thf, bis 2-di-tert-butylphosphino ethyl amine solution, 10 wt. % in thf
MJYKYLNPIGDVEF-UHFFFAOYSA-N
2-ditert-butylphosphanyl-N-(2-ditert-butylphosphanylethyl)ethanamine
51346231
10% w/w soln. in THF
0.911
C20H45NP2
([(CH3)3C]2PCH2CH2)2NH
1 g
Air Sensitive
Insoluble in water.
CC(C)(C)P(CCNCCP(C(C)(C)C)C(C)(C)C)C(C)(C)C
361.54
361.53
Bis[2-(di-tert-butylphosphino)ethyl]amine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H351-H315-H319-H335
Highly flammable liquid and vapor.
Suspected of causing cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P201-P202-P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P281-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P310-P330-P332+P313-P363-P370+P378q-P501c

H225-H302-H315-H318-H335-H336-H351

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures; Store under Argon

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only