Learn More
Bis(2-ethylhexyl)amine, 99%
CAS: 106-20-7 | C16H35N | 241.463 g/mol
$183.59 - $183.59
Chemical Identifiers
| CAS | 106-20-7 |
|---|---|
| Molecular Formula | C16H35N |
| Molecular Weight (g/mol) | 241.463 |
| MDL Number | MFCD00009489 |
| InChI Key | SAIKULLUBZKPDA-UHFFFAOYSA-N |
| Synonym | bis 2-ethylhexyl amine, di 2-ethylhexyl amine, diisooctylamine, 1-hexanamine, 2-ethyl-n-2-ethylhexyl, bis-2-ethylhexylamine, 2,2'-diethyldihexylamine, 2,2'-diethylhexylamine, di-2-ethylhexyl amine, dihexylamine, 2,2'-diethyl, ccris 4619 |
| PubChem CID | 7791 |
| IUPAC Name | 2-ethyl-N-(2-ethylhexyl)hexan-1-amine |
| SMILES | CCCCC(CC)CNCC(CC)CCCC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL13894AP
|
Thermo Scientific Chemicals
L13894AP |
500 mL |
Each for $183.59
|
|
|||||
Description
Bis(2-ethylhexyl)amine (BEA) was used in the synthesis of stable nanosized CdS particles coated by an oriented monolayer of chemically bonded BEA molecules
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsBis(2-ethylhexyl)amine (BEA) was used in the synthesis of stable nanosized CdS particles coated by an oriented monolayer of chemically bonded BEA molecules
Solubility
Soluble in water 0.045 g/L @ 20ËC.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
Chemical Identifiers
| 106-20-7 | |
| 241.463 | |
| SAIKULLUBZKPDA-UHFFFAOYSA-N | |
| 7791 | |
| CCCCC(CC)CNCC(CC)CCCC |
| C16H35N | |
| MFCD00009489 | |
| bis 2-ethylhexyl amine, di 2-ethylhexyl amine, diisooctylamine, 1-hexanamine, 2-ethyl-n-2-ethylhexyl, bis-2-ethylhexylamine, 2,2'-diethyldihexylamine, 2,2'-diethylhexylamine, di-2-ethylhexyl amine, dihexylamine, 2,2'-diethyl, ccris 4619 | |
| 2-ethyl-N-(2-ethylhexyl)hexan-1-amine |
Specifications
| 106-20-7 | |
| 0.805 | |
| 121°C (250°F) | |
| 1.442 | |
| 500 mL | |
| 1748342 | |
| Soluble in water 0.045g/L @ 20?C. | |
| CCCCC(CC)CNCC(CC)CCCC | |
| 241.463 | |
| 241.46 | |
| Bis(2-ethylhexyl)amine |
| <-60°C | |
| 281°C | |
| C16H35N | |
| MFCD00009489 | |
| UN2922 | |
| bis 2-ethylhexyl amine, di 2-ethylhexyl amine, diisooctylamine, 1-hexanamine, 2-ethyl-n-2-ethylhexyl, bis-2-ethylhexylamine, 2,2'-diethyldihexylamine, 2,2'-diethylhexylamine, di-2-ethylhexyl amine, dihexylamine, 2,2'-diethyl, ccris 4619 | |
| SAIKULLUBZKPDA-UHFFFAOYSA-N | |
| 2-ethyl-N-(2-ethylhexyl)hexan-1-amine | |
| 7791 | |
| 99% |
Safety and Handling
GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302-H311+H331-H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
EINECSNumber : 203-372-4
RTECSNumber : IH6825000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only