missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Bis(2-ethylhexyl) terephthalate, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 402492500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Bis(2-ethylhexyl) terephthalate | |
6422-86-2 | |
100.0 | |
Colorless | |
400.0°C | |
Authentic | |
Glass bottle | |
1.4890 to 1.4910 | |
250 g | |
0.984 | |
Solubility in water: insoluble | |
CCCCC(CC)COC(=O)C1=CC=C(C=C1)C(=O)OCC(CC)CCCC | |
390.55 | |
390.55 | |
Liquid |
97% | |
96 | |
-67.0°C | |
0.9840g/mL | |
>110°C | |
96% min. (GC) | |
C24H38O4 | |
MFCD00072256 | |
09, IV, 3306 | |
bis 2-ethylhexyl terephthalate, kodaflex dotp, di-2-ethylhexyl terephthalate, 1,4-benzenedicarboxylic acid, bis 2-ethylhexyl ester, terephthalic acid, bis 2-ethylhexyl ester, eastman 168, ccris 7052, di 2-ethylhexyl terephthalate, bis 2-ethylhexyl benzene-1,4-dicarboxylate, 1,4-benzenedicarboxylic acid, 1,4-bis 2-ethylhexyl ester | |
RWPICVVBGZBXNA-UHFFFAOYSA-N | |
bis(2-ethylhexyl) benzene-1,4-dicarboxylate | |
22932 | |
97% |
Chemical Identifiers
6422-86-2 | |
390.55 | |
RWPICVVBGZBXNA-UHFFFAOYSA-N | |
22932 | |
CCCCC(CC)COC(=O)C1=CC=C(C=C1)C(=O)OCC(CC)CCCC |
C24H38O4 | |
MFCD00072256 | |
bis 2-ethylhexyl terephthalate, kodaflex dotp, di-2-ethylhexyl terephthalate, 1,4-benzenedicarboxylic acid, bis 2-ethylhexyl ester, terephthalic acid, bis 2-ethylhexyl ester, eastman 168, ccris 7052, di 2-ethylhexyl terephthalate, bis 2-ethylhexyl benzene-1,4-dicarboxylate, 1,4-benzenedicarboxylic acid, 1,4-bis 2-ethylhexyl ester | |
bis(2-ethylhexyl) benzene-1,4-dicarboxylate |
Safety and Handling
EINECSNumber : 229-176-9
RTECSNumber : WZ0883500
TSCA : TSCA
RUO – Research Use Only