missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diethylene glycol, 99%, extra pure, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 121160010

 View more versions of this product

Catalog No. AC121160010



Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Diethylene glycol
98.5
-10.0°C
245.0°C
Authentic
Glass bottle
1.4450 to 1.4480
MFCD00002882
01, 468
15, 3131
Solubility in water: soluble. Other solubilities: soluble in ethanol, acetone, ether,, ethylene glycol, insoluble in benzene, toluene, carbon, tetrachloride and aliphatic hydrocarbons
OCCOCCO
106.12
38 mPa.s (20°C)
106.12
Extra Pure
111-46-6
100.0
1.1100g/mL
143°C
98.5% min. (GC)
C4H10O3
(HOCH2CH2)2O
1 L
1.11
diethylene glycol, 2,2'-oxydiethanol, diglycol, diethylenglykol, 2-hydroxyethyl ether, bis 2-hydroxyethyl ether, ethanol, 2,2'-oxybis, 2,2'-oxybisethanol, 2-2-hydroxyethoxy ethanol, digol
MTHSVFCYNBDYFN-UHFFFAOYSA-N
2-(2-hydroxyethoxy)ethan-1-ol
8117
CHEBI:46807
99%
Oily Liquid

Chemical Identifiers

111-46-6
106.12
MTHSVFCYNBDYFN-UHFFFAOYSA-N
8117
OCCOCCO
C4H10O3
MFCD00002882
diethylene glycol, 2,2'-oxydiethanol, diglycol, diethylenglykol, 2-hydroxyethyl ether, bis 2-hydroxyethyl ether, ethanol, 2,2'-oxybis, 2,2'-oxybisethanol, 2-2-hydroxyethoxy ethanol, digol
CHEBI:46807
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
May cause damage to organs through prolonged or repeated exposure.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

GHS Signal Word: Warning

EINECSNumber : 203-872-2

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only