missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bis(2-hydroxyethyl) Terephthalate 85.0+%, TCI America™

$59.25 - $286.38

Chemical Identifiers

CAS 959-26-2
Molecular Formula C12H14O6
Molecular Weight (g/mol) 254.238
MDL Number MFCD00081066
InChI Key QPKOBORKPHRBPS-UHFFFAOYSA-N
Synonym Terephthalic Acid Bis(2-hydroxyethyl) Ester, BHET
PubChem CID 13739
IUPAC Name bis(2-hydroxyethyl) benzene-1,4-dicarboxylate
SMILES C1=CC(=CC=C1C(=O)OCCO)C(=O)OCCO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B342925G
View Documents
TCI America
B342925G
25 g
Each for $59.25
 
B3429500G
View Documents
TCI America
B3429500G
500 g
Each for $286.38
 
Description

Description

Specifications

Chemical Identifiers

959-26-2
254.238
QPKOBORKPHRBPS-UHFFFAOYSA-N
13739
C1=CC(=CC=C1C(=O)OCCO)C(=O)OCCO
C12H14O6
MFCD00081066
Terephthalic Acid Bis(2-hydroxyethyl) Ester, BHET
bis(2-hydroxyethyl) benzene-1,4-dicarboxylate

Specifications

959-26-2
White
MFCD00081066
Terephthalic Acid Bis(2-hydroxyethyl) Ester, BHET
C1=CC(=CC=C1C(=O)OCCO)C(=O)OCCO
254.238
254.24
Crystalline Powder
106°C
C12H14O6
25 g
QPKOBORKPHRBPS-UHFFFAOYSA-N
bis(2-hydroxyethyl) benzene-1,4-dicarboxylate
13739
≥85.0% (HPLC)
Bis(2-hydroxyethyl) Terephthalate
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-1322&(3)-1327

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'