missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
[Bis(2-methoxyethyl)amino]sulfur trifluoride, 2.7M (50 wt.%) solution in toluene, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 368960050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagentSpecifications
[Bis(2-methoxyethyl)amino]sulfur trifluoride | |
49.0,49.0 | |
Orange-Yellow | |
4°C | |
C6H14F3NO2S | |
5 g | |
15,2899 | |
APOYTRAZFJURPB-UHFFFAOYSA-N | |
2-methoxy-N-(2-methoxyethyl)-N-(trifluoro-$l^{4}-sulfanyl)ethanamine | |
2734690 | |
≥95% (NMR, neat substance) |
202289-38-1,108-88-3 | |
51.0,51.0 | |
1.2000g/mL | |
Plastic bottle | |
(CH3OCHCH2)2NSF3 | |
1.2 | |
bis 2-methoxyethyl aminosulfur trifluoride, deoxo-fluor, 2-methoxy-n-2-methoxyethyl-n-trifluorothio-ethanamine, deoxo-fluor r, bis 2-methoxyethyl aminosulphurtrifluoride, bast, bis 2-methoxyethyl amino sulfur trifluoride, bis 2-methoxyethyl aminosulphur trifluoride | |
COCCN(CCOC)S(F)(F)F | |
221.24 | |
221.24 | |
Liquid |
RUO – Research Use Only