missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Bis(3-methyl-1-phenyl-5-pyrazolone) 97.0+%, TCI America™
Supplier: TCI America B050825G
Specifications
Bis(3-methyl-1-phenyl-5-pyrazolone) [for Determination of CN, Vitamin B12, etc] | |
White-Yellow | |
MFCD00020765 | |
bispyrazolone, 4,4'-bis 1-phenyl-3-methyl-5-pyrazolone, 4,4'-bi-3h-pyrazole-3,3'-dione, 2,2',4,4'-tetrahydro-5,5'-dimethyl-2,2'-diphenyl, 4,4'-bis 3-methyl-1-phenyl-5-pyrazolone, 4,4'-bi-2-pyrazoline-5,5'-dione, 3,3'-dimethyl-1,1'-diphenyl, 3,3'-dimethyl-1,1'-diphenyl-1h,1'h-4,4'-bipyrazole-5,5' 4h,4'h-dione, 5-methyl-4-3-methyl-5-oxo-1-phenyl-4h-pyrazol-4-yl-2-phenyl-4h-pyrazol-3-one, 4,5'-dione, 3,3'-dimethyl-1,1'-diphenyl | |
CC1=NN(C(=O)C1C2C(=NN(C2=O)C3=CC=CC=C3)C)C4=CC=CC=C4 | |
346.39 | |
346.39 | |
Crystalline Powder |
7477-67-0 | |
C20H18N4O2 | |
25 g | |
FORCWSNQDMPPOC-UHFFFAOYSA-N | |
5-methyl-4-(3-methyl-5-oxo-1-phenyl-4H-pyrazol-4-yl)-2-phenyl-4H-pyrazol-3-one | |
95807 | |
≥97.0% (T) |
Chemical Identifiers
7477-67-0 | |
346.39 | |
FORCWSNQDMPPOC-UHFFFAOYSA-N | |
95807 | |
CC1=NN(C(=O)C1C2C(=NN(C2=O)C3=CC=CC=C3)C)C4=CC=CC=C4 |
C20H18N4O2 | |
MFCD00020765 | |
bispyrazolone, 4,4'-bis 1-phenyl-3-methyl-5-pyrazolone, 4,4'-bi-3h-pyrazole-3,3'-dione, 2,2',4,4'-tetrahydro-5,5'-dimethyl-2,2'-diphenyl, 4,4'-bis 3-methyl-1-phenyl-5-pyrazolone, 4,4'-bi-2-pyrazoline-5,5'-dione, 3,3'-dimethyl-1,1'-diphenyl, 3,3'-dimethyl-1,1'-diphenyl-1h,1'h-4,4'-bipyrazole-5,5' 4h,4'h-dione, 5-methyl-4-3-methyl-5-oxo-1-phenyl-4h-pyrazol-4-yl-2-phenyl-4h-pyrazol-3-one, 4,5'-dione, 3,3'-dimethyl-1,1'-diphenyl | |
5-methyl-4-(3-methyl-5-oxo-1-phenyl-4H-pyrazol-4-yl)-2-phenyl-4H-pyrazol-3-one |
Safety and Handling
TSCA : Yes