missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Bis(3-nitrophenyl) Disulfide 98.0+%, TCI America™
Supplier: TCI America B292925G
Specifications
Bis(3-nitrophenyl) Disulfide | |
83°C | |
MFCD00007246 | |
bis 3-nitrophenyl disulfide, nitrophenide, disulfide, bis 3-nitrophenyl, hinagen, 3-nitrophenyl disulfide, 3,3'-dinitrodiphenyl disulfide, megasul, bis m-nitrophenyl disulfide, m,m'-dinitrodiphenyl disulfide, 1-nitro-3-3-nitrophenyl disulfanyl benzene | |
C1=CC(=CC(=C1)SSC2=CC=CC(=C2)[N+](=O)[O-])[N+](=O)[O-] | |
308.326 | |
308.33 | |
Crystalline Powder |
537-91-7 | |
C12H8N2O4S2 | |
25 g | |
ODOFDWDUSSFUMN-UHFFFAOYSA-N | |
1-nitro-3-[(3-nitrophenyl)disulfanyl]benzene | |
10842 | |
≥98.0% (HPLC) |
Chemical Identifiers
537-91-7 | |
308.326 | |
ODOFDWDUSSFUMN-UHFFFAOYSA-N | |
10842 | |
C1=CC(=CC(=C1)SSC2=CC=CC(=C2)[N+](=O)[O-])[N+](=O)[O-] |
C12H8N2O4S2 | |
MFCD00007246 | |
bis 3-nitrophenyl disulfide, nitrophenide, disulfide, bis 3-nitrophenyl, hinagen, 3-nitrophenyl disulfide, 3,3'-dinitrodiphenyl disulfide, megasul, bis m-nitrophenyl disulfide, m,m'-dinitrodiphenyl disulfide, 1-nitro-3-3-nitrophenyl disulfanyl benzene | |
1-nitro-3-[(3-nitrophenyl)disulfanyl]benzene |
Safety and Handling
TSCA : Yes