missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Bis(4-aminophenyl) Sulfone 98.0+%, TCI America™
Supplier: TCI America D0089100G
Specifications
Bis(4-aminophenyl) Sulfone | |
177°C | |
MFCD00007887 | |
3077 | |
MQJKPEGWNLWLTK-UHFFFAOYSA-N | |
4-(4-aminophenyl)sulfonylaniline | |
2955 | |
248.30 | |
Crystalline Powder |
80-08-0 | |
C12H12N2O2S | |
100 g | |
dapsone, diaphenylsulfone, 4,4'-diaminodiphenyl sulfone, dapson, 4,4'-sulfonyldianiline, diaphenylsulfon, sulfona, 4-aminophenyl sulfone, avlosulfone, diphenasone | |
C1=CC(=CC=C1N)S(=O)(=O)C2=CC=C(C=C2)N | |
248.3 | |
CHEBI:4325 | |
≥98.0% (HPLC,T) |
Chemical Identifiers
80-08-0 | |
248.3 | |
MQJKPEGWNLWLTK-UHFFFAOYSA-N | |
2955 | |
4-(4-aminophenyl)sulfonylaniline |
C12H12N2O2S | |
MFCD00007887 | |
dapsone, diaphenylsulfone, 4,4'-diaminodiphenyl sulfone, dapson, 4,4'-sulfonyldianiline, diaphenylsulfon, sulfona, 4-aminophenyl sulfone, avlosulfone, diphenasone | |
CHEBI:4325 | |
C1=CC(=CC=C1N)S(=O)(=O)C2=CC=C(C=C2)N |
Safety and Handling
EINECSNumber : (3)-2163
RTECSNumber : BY8925000
TSCA : Yes