missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Bis(4-bromophenyl)diphenylsilane 98.0+%, TCI America™
$276.63 - $955.35
Chemical Identifiers
CAS | 18733-91-0 |
---|---|
Molecular Formula | C24H18Br2Si |
Molecular Weight (g/mol) | 494.301 |
InChI Key | UTEBJTKMYMQRIF-UHFFFAOYSA-N |
PubChem CID | 297512 |
IUPAC Name | bis(4-bromophenyl)-diphenylsilane |
SMILES | C1=CC=C(C=C1)[Si](C2=CC=CC=C2)(C3=CC=C(C=C3)Br)C4=CC=C(C=C4)Br |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
B5087200MG
|
TCI America
B5087200MG |
200 mg |
Each for $276.63
|
|
|||||
B50871G
|
TCI America
B50871G |
1 g |
Each for $955.35
|
|
|||||
Chemical Identifiers
18733-91-0 | |
494.301 | |
297512 | |
C1=CC=C(C=C1)[Si](C2=CC=CC=C2)(C3=CC=C(C=C3)Br)C4=CC=C(C=C4)Br |
C24H18Br2Si | |
UTEBJTKMYMQRIF-UHFFFAOYSA-N | |
bis(4-bromophenyl)-diphenylsilane |
Specifications
18733-91-0 | |
White | |
200 mg | |
C1=CC=C(C=C1)[Si](C2=CC=CC=C2)(C3=CC=C(C=C3)Br)C4=CC=C(C=C4)Br | |
494.301 | |
494.30 | |
Crystalline Powder |
169°C | |
C24H18Br2Si | |
UTEBJTKMYMQRIF-UHFFFAOYSA-N | |
bis(4-bromophenyl)-diphenylsilane | |
297512 | |
≥98.0% (GC) | |
Bis(4-bromophenyl)diphenylsilane |
Safety and Handling
TSCA : No