Learn More
Bis(4-fluorobenzyl)amine, 97%, Thermo Scientific Chemicals
$248.36 - $248.36
Chemical Identifiers
CAS | 134227-41-1 |
---|---|
Molecular Formula | C14H13F2N |
Molecular Weight (g/mol) | 233.262 |
MDL Number | MFCD05155916 |
InChI Key | YKFNPVWWPPJNAQ-UHFFFAOYSA-N |
Synonym | bis 4-fluorobenzyl amine, bis 4-fluorophenyl methyl amine, bis p-fluorobenzyl amine, di-4-fluorobenzyl amine, bis-4-fluoro-benzyl-amine, n,n-bis 4-fluorobenzyl amine, 1-4-fluorophenyl-n-4-fluorophenyl methyl methanamine |
PubChem CID | 834401 |
IUPAC Name | 1-(4-fluorophenyl)-N-[(4-fluorophenyl)methyl]methanamine |
SMILES | C1=CC(=CC=C1CNCC2=CC=C(C=C2)F)F |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH56467MD
|
Thermo Scientific Chemicals
H56467MD |
250 mg |
Each for $248.36
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
134227-41-1 | |
233.262 | |
YKFNPVWWPPJNAQ-UHFFFAOYSA-N | |
834401 | |
C1=CC(=CC=C1CNCC2=CC=C(C=C2)F)F |
C14H13F2N | |
MFCD05155916 | |
bis 4-fluorobenzyl amine, bis 4-fluorophenyl methyl amine, bis p-fluorobenzyl amine, di-4-fluorobenzyl amine, bis-4-fluoro-benzyl-amine, n,n-bis 4-fluorobenzyl amine, 1-4-fluorophenyl-n-4-fluorophenyl methyl methanamine | |
1-(4-fluorophenyl)-N-[(4-fluorophenyl)methyl]methanamine |
Specifications
134227-41-1 | |
MFCD05155916 | |
UN3259 | |
YKFNPVWWPPJNAQ-UHFFFAOYSA-N | |
1-(4-fluorophenyl)-N-[(4-fluorophenyl)methyl]methanamine | |
834401 | |
97% |
C14H13F2N | |
250 mg | |
bis 4-fluorobenzyl amine, bis 4-fluorophenyl methyl amine, bis p-fluorobenzyl amine, di-4-fluorobenzyl amine, bis-4-fluoro-benzyl-amine, n,n-bis 4-fluorobenzyl amine, 1-4-fluorophenyl-n-4-fluorophenyl methyl methanamine | |
C1=CC(=CC=C1CNCC2=CC=C(C=C2)F)F | |
233.262 | |
233.26 | |
Bis(4-fluorobenzyl)amine |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only