missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Bis(4-methoxyphenyl)disulfide, 97%, Thermo Scientific Chemicals
$107.85 - $350.21
Chemical Identifiers
CAS | 5335-87-5 |
---|---|
Molecular Formula | C14H14O2S2 |
Molecular Weight (g/mol) | 278.38 |
MDL Number | MFCD00041358 |
InChI Key | PZQGLCGLPMWYBT-UHFFFAOYSA-N |
Synonym | bis 4-methoxyphenyl disulfide, disulfide, bis 4-methoxyphenyl, p-methoxyphenyl disulfide, 4,4'-dimethoxy diphenyldisulfide, 4-methoxyphenyl disulfide, anisole, 4,4'-dithiodi, 1-methoxy-4-4-methoxyphenyl disulfanyl benzene, 1,2-bis 4-methoxyphenyl disulfane, bis p-methoxyphenyl disulfide |
PubChem CID | 79258 |
IUPAC Name | 1-methoxy-4-[(4-methoxyphenyl)disulfanyl]benzene |
SMILES | COC1=CC=C(SSC2=CC=C(OC)C=C2)C=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2539903
|
Thermo Scientific Chemicals
B2539903 |
1 g |
Each for $107.85
|
|
|||||
AAB2539906
|
Thermo Scientific Chemicals
B2539906 |
5 g |
Each for $350.21
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
5335-87-5 | |
278.38 | |
PZQGLCGLPMWYBT-UHFFFAOYSA-N | |
79258 | |
COC1=CC=C(SSC2=CC=C(OC)C=C2)C=C1 |
C14H14O2S2 | |
MFCD00041358 | |
bis 4-methoxyphenyl disulfide, disulfide, bis 4-methoxyphenyl, p-methoxyphenyl disulfide, 4,4'-dimethoxy diphenyldisulfide, 4-methoxyphenyl disulfide, anisole, 4,4'-dithiodi, 1-methoxy-4-4-methoxyphenyl disulfanyl benzene, 1,2-bis 4-methoxyphenyl disulfane, bis p-methoxyphenyl disulfide | |
1-methoxy-4-[(4-methoxyphenyl)disulfanyl]benzene |
Specifications
5335-87-5 | |
200°C (3 mmHg) | |
MFCD00041358 | |
1914789 | |
PZQGLCGLPMWYBT-UHFFFAOYSA-N | |
1-methoxy-4-[(4-methoxyphenyl)disulfanyl]benzene | |
79258 | |
97% |
37°C to 41°C | |
C14H14O2S2 | |
1 g | |
bis 4-methoxyphenyl disulfide, disulfide, bis 4-methoxyphenyl, p-methoxyphenyl disulfide, 4,4'-dimethoxy diphenyldisulfide, 4-methoxyphenyl disulfide, anisole, 4,4'-dithiodi, 1-methoxy-4-4-methoxyphenyl disulfanyl benzene, 1,2-bis 4-methoxyphenyl disulfane, bis p-methoxyphenyl disulfide | |
COC1=CC=C(SSC2=CC=C(OC)C=C2)C=C1 | |
278.38 | |
278.39 | |
Bis(4-methoxyphenyl)disulfide |
Safety and Handling
GHS H Statement
H319
Causes serious eye irritation.
P264b-P280i-P305+P351+P338
H319
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only