missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Bis(4-tert-butylphenyl)iodonium p-toluenesulfonate, Electronic grade, ≥99% (metals basis), Thermo Scientific Chemicals
$200.88 - $909.44
Chemical Identifiers
CAS | 131717-99-2 |
---|---|
Molecular Formula | C27H33IO3S |
Molecular Weight (g/mol) | 564.52 |
MDL Number | MFCD02683472 |
InChI Key | UEJFJTOGXLEPIV-UHFFFAOYSA-M |
Synonym | bis 4-tert-butylphenyl iodanium tosylate, bis 4-tert-butylphenyl iodonium p-toluenesulfonate, 4,4-di-tert-butyldiphenyliodonium p-toluenesulfonate, acmc-20n1fr, bis 4-tert-butylphenyl iodonium tosylate, bis 4-tert-butylphenl iodonium p, bis p-tert-butylphenyl iodonium p-toluenesulfonate, bis 4-tert-butylphenyl iodonium p-toluenesulfonate, electronic grade, bis 4-tert-butylphenyl iodonium p-toluenesulfonate, electronic grade trace metals basis |
PubChem CID | 16216932 |
IUPAC Name | bis(4-tert-butylphenyl)iodanium;4-methylbenzenesulfonate |
SMILES | CC1=CC=C(C=C1)S([O-])(=O)=O.CC(C)(C)C1=CC=C([I+]C2=CC=C(C=C2)C(C)(C)C)C=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAH5518803
|
Thermo Scientific Chemicals
H5518803 |
1 g |
N/A
|
|
||||||
AAH5518806
|
Thermo Scientific Chemicals
H5518806 |
5 g |
N/A
|
|
||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
131717-99-2 | |
564.52 | |
UEJFJTOGXLEPIV-UHFFFAOYSA-M | |
16216932 | |
CC1=CC=C(C=C1)S([O-])(=O)=O.CC(C)(C)C1=CC=C([I+]C2=CC=C(C=C2)C(C)(C)C)C=C1 |
C27H33IO3S | |
MFCD02683472 | |
bis 4-tert-butylphenyl iodanium tosylate, bis 4-tert-butylphenyl iodonium p-toluenesulfonate, 4,4-di-tert-butyldiphenyliodonium p-toluenesulfonate, acmc-20n1fr, bis 4-tert-butylphenyl iodonium tosylate, bis 4-tert-butylphenl iodonium p, bis p-tert-butylphenyl iodonium p-toluenesulfonate, bis 4-tert-butylphenyl iodonium p-toluenesulfonate, electronic grade, bis 4-tert-butylphenyl iodonium p-toluenesulfonate, electronic grade trace metals basis | |
bis(4-tert-butylphenyl)iodanium;4-methylbenzenesulfonate |
Specifications
131717-99-2 | |
White | |
MFCD02683472 | |
Light sensitive | |
UEJFJTOGXLEPIV-UHFFFAOYSA-M | |
bis(4-tert-butylphenyl)iodanium;4-methylbenzenesulfonate | |
16216932 | |
≥99% | |
Powder | |
(metals basis) |
237°C to 240°C | |
C27H33IO3S | |
1 g | |
bis 4-tert-butylphenyl iodanium tosylate, bis 4-tert-butylphenyl iodonium p-toluenesulfonate, 4,4-di-tert-butyldiphenyliodonium p-toluenesulfonate, acmc-20n1fr, bis 4-tert-butylphenyl iodonium tosylate, bis 4-tert-butylphenl iodonium p, bis p-tert-butylphenyl iodonium p-toluenesulfonate, bis 4-tert-butylphenyl iodonium p-toluenesulfonate, electronic grade, bis 4-tert-butylphenyl iodonium p-toluenesulfonate, electronic grade trace metals basis | |
CC1=CC=C(C=C1)S([O-])(=O)=O.CC(C)(C)C1=CC=C([I+]C2=CC=C(C=C2)C(C)(C)C)C=C1 | |
564.52 | |
564.52 | |
Electronic | |
Bis(4-tert-butylphenyl)iodonium p-toluenesulfonate |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only