missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bis(8-quinolinolato)copper(II) 95.0+%, TCI America™

Supplier:  TCI America B167725G

Catalog No. B167725G

Explore available promotions

Description

Description

Specifications

Specifications

Bis(8-quinolinolato)copper(II)
Green-Yellow
MFCD00067392
3077
YXLXNENXOJSQEI-UHFFFAOYSA-L
copper(2+) bis(quinolin-8-olate)
56840841
351.85
Crystalline Powder
10380-28-6
C18H12CuN2O2
25 g
cunilate, cuproquin, dokirin, fruitdo, quinolate, quinondo, milmer, copper oxinate, copper oxine, copper-oxine
[Cu++].[O-]C1=C2N=CC=CC2=CC=C1.[O-]C1=C2N=CC=CC2=CC=C1
351.85
CHEBI:82028
≥95.0% (T)

Chemical Identifiers

10380-28-6
351.85
YXLXNENXOJSQEI-UHFFFAOYSA-L
56840841
copper(2+) bis(quinolin-8-olate)
C18H12CuN2O2
MFCD00067392
cunilate, cuproquin, dokirin, fruitdo, quinolate, quinondo, milmer, copper oxinate, copper oxine, copper-oxine
CHEBI:82028
[Cu++].[O-]C1=C2N=CC=CC2=CC=C1.[O-]C1=C2N=CC=CC2=CC=C1
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (5)-0805

RTECSNumber : VC5250000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions