missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Bis(aminomethyl)norbornane (mixture of isomers) 98.0+%, TCI America™
$71.10 - $290.32
Chemical Identifiers
CAS | 56602-77-8 |
---|---|
Molecular Formula | C9H18N2 |
Molecular Weight (g/mol) | 154.26 |
MDL Number | MFCD19705260 |
InChI Key | RPYFJVIASOJLJS-UHFFFAOYNA-N |
Synonym | Bicyclo[2.2.1]heptanedimethanamine, Bis(aminomethyl)bicyclo[2.2.1]heptane |
PubChem CID | 10857171 |
IUPAC Name | 1-[3-(aminomethyl)bicyclo[2.2.1]heptan-2-yl]methanamine |
SMILES | NCC1C2CCC(C2)C1CN |
Chemical Identifiers
56602-77-8 | |
154.26 | |
RPYFJVIASOJLJS-UHFFFAOYNA-N | |
10857171 | |
NCC1C2CCC(C2)C1CN |
C9H18N2 | |
MFCD19705260 | |
Bicyclo[2.2.1]heptanedimethanamine, Bis(aminomethyl)bicyclo[2.2.1]heptane | |
1-[3-(aminomethyl)bicyclo[2.2.1]heptan-2-yl]methanamine |
Specifications
56602-77-8 | |
C9H18N2 | |
25 g | |
Bicyclo[2.2.1]heptanedimethanamine, Bis(aminomethyl)bicyclo[2.2.1]heptane | |
NCC1C2CCC(C2)C1CN | |
154.26 | |
154.26 | |
Liquid |
Colorless | |
MFCD19705260 | |
2735 | |
RPYFJVIASOJLJS-UHFFFAOYNA-N | |
1-[3-(aminomethyl)bicyclo[2.2.1]heptan-2-yl]methanamine | |
10857171 | |
≥98.0% (T) | |
Bis(aminomethyl)norbornane (mixture of isomers) |
Safety and Handling
EINECSNumber : (4)-1732
TSCA : Yes